Learn More
Enzo Life Sciences TRAIL-R2 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ALX210743C200
Encompass_Preferred
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 68-94 (A68PQQKRSSPSEGL CPPGHHISEDGRDC94) of the extracellular domain of human TRAIL-R2. Species reactivity: Human, Mouse. Applications: Flow Cytometry, ICC, IHC (PS), WB. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8147