Learn More
Medchemexpress LLC TRAIL R2/TNFRSF10B 10ug
SDP
Catalog No. 5000180185
Encompass_Preferred
TRAIL R2/TNFRSF10B Protein Human (HEK293) is a cell surface receptor of the TNF-receptor superfamily that binds TRAIL and mediates apoptosis
Catalog No. 50-001-80185 Supplier Medchemexpress LLC Supplier No. HYP730710UG
TRAIL R2/TNFRSF10B Protein Human (HEK293) is a cell surface receptor of the TNF-receptor superfamily that binds TRAIL and mediates apoptosis