Learn More
Enzo Life Sciences TRAIL-R3 (human) monoclonal antibody (DJR3) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX8049080100
Encompass_Preferred
Clone: DJR3. Isotype: IgG1. Immunogen: Recombinant human TRAIL-R3 extracellular domain IgG1 Fc fusion protein. Species reactivity: Human. Applications: Flow Cytometry. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-0019