Learn More
Enzo Life Sciences TRAIL-R4 (human) monoclonal antibody (01) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX8049150100
Encompass_Preferred
Clone: 01. Isotype: IgG1. Immunogen: Recombinant human TRAIL-R4 (aa 1-210) IgG1 Fc fusion protein. Species reactivity: Human. Applications: Flow Cytometry, IHC (PS). Formulation: Liquid. In PBS, pH 7.4.  Contains no preservatives.
Catalog No. 50-201-0023