Learn More
Enzo Life Sciences TRAIL-R4 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP371E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human TRAIL-R4. Species reactivity: Human, Mouse, Rat. Applications: ICC, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7163