Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TRAIL R4/ TNFRSF10D ELISA Kit

Catalog No. EHTNFRSF10D
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHTNFRSF10D 96 Tests
50-274-1190 5 x 96 Tests
50-255-0904 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHTNFRSF10D Supplier Invitrogen™ Supplier No. EHTNFRSF10D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human TRAIL R4 TNFRSF10D (TRAIL R4) ELISA quantitates Hu TRAIL R4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TRAIL R4. Principle of the method The Human TRAIL R4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. A fourth member of the emerging TRAIL receptor family, TRAIL-R4, has signaling capacity similar to that of TRAIL-R1 and TRAIL-R2 with respect to NF-kappaB activation, but differs in its inability to induce apoptosis. Like TRAIL-R1 and TRAIL-R2, this receptor shows widespread tissue expression. The human TRAIL-R4 gene has been mapped to chromosome 8p22-21, clustered with three other TRAIL receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBN6
Assay Range 10-4000 pg/mL
Assay Sensitivity 10 pg/mL
Calibration Range 10 pg/mL to 4000 pg/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CD264; DCR2; Decoy receptor 2; TNF receptor superfamily member 10d; TNF receptor-related receptor for TRAIL; TNF-related apoptosis-inducing ligand receptor 4; TNFRSF10D; TRAIL receptor 4; TRAIL receptor with a truncated death domain; TRAILR4; TRAIL-R4; TRUNDD; Tumor necrosis factor receptor superfamily member 10D; tumor necrosis factor receptor superfamily member 10d, decoy with truncated death domain; tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain; UNQ251/PRO288CD264; DCR2; Decoy receptor 2; TNF receptor superfamily member 10d; TNF receptor-related receptor for TRAIL; TNF-related apoptosis-inducing ligand receptor 4; TNFRSF10D; TRAIL receptor 4; TRAIL receptor with a truncated death domain; TRAILR4; TRAIL-R4; TRUNDD; Tumor necrosis factor receptor superfamily member 10D; tumor necrosis factor receptor superfamily member 10d, decoy with truncated death domain; tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain; UNQ251/PRO288; CD264; DCR2; TRAIL-R4; TRAILR4; TRUNDD
Gene ID (Entrez) 8793
Gene Symbol TNFRSF10D
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Apoptosis
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym UNQ251/PRO288, CD264, DCR2, TRAIL-R4, TRAILR4, TRUNDD
Target TRAIL-R4 (CD264)
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.