Learn More
MSC Honeywell Safety MTS Trail Safety Shoes, Size 13.5 US, Composite Toe, S3S Rated, 1 Pair
Catalog No. 5000507232
Encompass
The MTS Trail safety shoe is a lightweight, non-metallic footwear option designed for a sporty, casual aesthetic suitable for indoor and outdoor environments. Engineered for use in logistics, services, transportation, and warehousing, this shoe combines comfort with essential protection.
  • Composite toe cap provides reliable impact and compression protection
  • Rated S3S SR FO HI CI ESD per EN ISO 20345:2022 standards
  • Anti-static and ESD-compliant construction
  • Features slip, oil, heat, and cold resistance
  • Puncture-resistant midsole utilizing non-metallic material
  • Includes heel energy absorption and a supportive TPU heel counter
  • Upper constructed from durable Ondura Nilo and Onsuede
  • Secured with round tubular laces and plastic hooks
  • Compliant with DGUV 112-191 for use with orthopedic insoles
Catalog No. 50-005-07232 Supplier MSC Supplier No. 79493359
The MTS Trail safety shoe is a lightweight, non-metallic footwear option designed for a sporty, casual aesthetic suitable for indoor and outdoor environments. Engineered for use in logistics, services, transportation, and warehousing, this shoe combines comfort with essential protection.
  • Composite toe cap provides reliable impact and compression protection
  • Rated S3S SR FO HI CI ESD per EN ISO 20345:2022 standards
  • Anti-static and ESD-compliant construction
  • Features slip, oil, heat, and cold resistance
  • Puncture-resistant midsole utilizing non-metallic material
  • Includes heel energy absorption and a supportive TPU heel counter
  • Upper constructed from durable Ondura Nilo and Onsuede
  • Secured with round tubular laces and plastic hooks
  • Compliant with DGUV 112-191 for use with orthopedic insoles