Learn More
Enzo Life Sciences TRAIL (soluble) (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX5220033010
Encompass_Preferred
Alternative name: Apo-2L, TNFSF 10, CD253. Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Specificity: Binds to human and mouse TRAIL receptors and human osteoprotegerin (OPG). Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9357