Learn More
Enzo Life Sciences TRAIL (soluble) (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE7210100
Encompass_Preferred
Alternative name: Apo-2L. TNFSF 10. CD253. MW: ~23kDa. Endotoxin content: Limulus amebocyte lysate (LAL) test. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In PBS, containing 10% glycerol and 0.1mM DTT. Source: Produced in E. coli. Human TRAIL (aa 114-281) is fused to a His-tag. Specificity: Binds to human and mouse TRAIL receptors and human osteoprotegerin (OPG). Use/Stability: When stored at -80°C, this protein is stable at the. Concentration supplied, in its current storage buffer. Procedures such as dilution of the protein, followed by refreezing, could lead to loss of activity. UniProt: P50591. Handling: After initial defrost, aliquot product into individual tubes and refreeze at -70°C. Avoid freeze/thaw cycles. Long Term Storage: -80°C
Catalog No. NC9964768