Learn More
Enzo Life Sciences TRAIL (soluble) (mouse), (recombinant) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE7220100
Encompass_Preferred
Alternative name: Apo-2L, TNFSF 10, CD253. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In PBS containing 0.1mM DTT. Source: Produced in E. coli. Soluble mouse TRAIL (aa 118-291). Long Term Storage: -80°C.
Catalog No. 50-201-2098