Learn More
Medchemexpress LLC TRAIL/TNFSF10 Trimer 100ug | 100ug
SDP
Catalog No. 5000194425
Encompass_Preferred
TRAIL Protein (TNFSF10) a member of the TNF superfamily is a type II transmembrane protein TRAIL Protein mainly interacts with TRAIL-R1 and TRAIL-R2 and induces apoptosis in tumor or infected cells TRAIL Protein also binds with DR4 DR5 and OPG TRAIL Protein can recruit FADD and further recruit and activates caspase-8 after binding to DR4 or DR5 Besides TRAIL may also trigger nonapoptotic signaling through activating pro-inflammatory pathways[1][2] TRAIL protein is mainly expressed on surface of immune cells such as cytotoxic T cells and natural killer (NK) cell[1] TRAIL/TNFSF10 Protein Human (HEK293 His-Flag) is a recombinant human TRAIL (G118-G281) with N-terminal His and Flag tag which is expressed in HEK293
Catalog No. 50-001-94425 Supplier Medchemexpress LLC Supplier No. HYP78528100UG
TRAIL Protein (TNFSF10) a member of the TNF superfamily is a type II transmembrane protein TRAIL Protein mainly interacts with TRAIL-R1 and TRAIL-R2 and induces apoptosis in tumor or infected cells TRAIL Protein also binds with DR4 DR5 and OPG TRAIL Protein can recruit FADD and further recruit and activates caspase-8 after binding to DR4 or DR5 Besides TRAIL may also trigger nonapoptotic signaling through activating pro-inflammatory pathways[1][2] TRAIL protein is mainly expressed on surface of immune cells such as cytotoxic T cells and natural killer (NK) cell[1] TRAIL/TNFSF10 Protein Human (HEK293 His-Flag) is a recombinant human TRAIL (G118-G281) with N-terminal His and Flag tag which is expressed in HEK293