Learn More
ACROBiosystems Human TRAIL R1 Protein, Fc Tag, 100 μg
Catalog No. 501657972
Encompass_Preferred
Human TRAIL R1, also known as TNFRSF10A, DR4, CD261, or APO2, is a receptor for the cytotoxic ligand TNFSF10 (TRAIL). This protein belongs to the TNF superfamily and contains one death domain and three TNFR-Cys repeats. Upon binding to its ligand, it recruits the adapter molecule FADD and caspase-8 to form the death-inducing signaling complex (DISC), which initiates the caspase cascade to mediate apoptosis. This protein is expressed in human HEK293 cells and features a human IgG1 Fc tag at the C-terminus.
  • Expressed in human HEK293 cells
  • Purity >95% as determined by SDS-PAGE
  • Calculated molecular weight of 49.0 kDa (migrates at 45–50 kDa due to glycosylation)
  • Endotoxin level less than 1.0 EU per μg
  • Formulated in 50 mM Tris, 100 mM Glycine, pH 7.5 with trehalose
Catalog No. 50-165-7972 Supplier ACROBiosystems Supplier No. TR1H5254100UG
Human TRAIL R1, also known as TNFRSF10A, DR4, CD261, or APO2, is a receptor for the cytotoxic ligand TNFSF10 (TRAIL). This protein belongs to the TNF superfamily and contains one death domain and three TNFR-Cys repeats. Upon binding to its ligand, it recruits the adapter molecule FADD and caspase-8 to form the death-inducing signaling complex (DISC), which initiates the caspase cascade to mediate apoptosis. This protein is expressed in human HEK293 cells and features a human IgG1 Fc tag at the C-terminus.
  • Expressed in human HEK293 cells
  • Purity >95% as determined by SDS-PAGE
  • Calculated molecular weight of 49.0 kDa (migrates at 45–50 kDa due to glycosylation)
  • Endotoxin level less than 1.0 EU per μg
  • Formulated in 50 mM Tris, 100 mM Glycine, pH 7.5 with trehalose