Learn More
ACROBiosystems Recombinant Protein;Human TRAIL R1 / DR4 / TNFRSF10A Protein;HEK293;1MG;TR1,TNFRSF10A,TRAILR1,CD261,APO2,DR4,TRAIL-R1,TRAILR-1,APO-2,DR-4
Catalog No. 501657896
Encompass_Preferred
Recombinant Protein;Human TRAIL R1 / DR4 / TNFRSF10A Protein;1MG;Human DR4, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession # NP_003835).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.0 kDa. The protein migrates as 19-23 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-7896 Supplier ACROBiosystems Supplier No. TR1H52281MG
Recombinant Protein;Human TRAIL R1 / DR4 / TNFRSF10A Protein;1MG;Human DR4, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession # NP_003835).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.0 kDa. The protein migrates as 19-23 kDa under reducing (R) condition (SDS-PAGE).