Learn More
ACROBiosystems ACROBiosystems Mouse TRAIL R2 / DR5 / TNFRSF10B Protein, His Tag
Catalog No. 501956916
Encompass_Preferred
Recombinant Protein;1MG;Mouse TRAIL R2, His Tag (TR2-M52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 53 - Lys 180 (Accession # Q9QZM4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.1 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TNFSF10 at 2 μg/mL (100 μL/well) can bind Mouse TRAIL R2, His Tag (Cat. No. TR2-M52H5) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-195-6916 Supplier ACROBiosystems Supplier No. TR2M52H51MG
Recombinant Protein;1MG;Mouse TRAIL R2, His Tag (TR2-M52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 53 - Lys 180 (Accession # Q9QZM4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.1 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TNFSF10 at 2 μg/mL (100 μL/well) can bind Mouse TRAIL R2, His Tag (Cat. No. TR2-M52H5) with a linear range of 0.039-0.625 μg/mL (QC tested).