Learn More
ACROBiosystems Recombinant Protein;Human TRAIL R2 / DR5 / TNFRSF10B Protein, Fc Tag;HEK293;100UG;TR2,TNFRSF10B,TRAILR2,TRAIL-R2,CD262,DR5,DR-5,KILLER,TRICK2,ZTNFR9,TRICK2A,TRICK2B,TRICKB,TRICK-2
Supplier:  ACROBiosystems TR2H5255100UG
Encompass_Preferred
Recombinant Protein;Human TRAIL R2 / DR5 / TNFRSF10B Protein, Fc Tag;100UG;Human DR5, Fc Tag (TR2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession # NP_003833).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. The protein migrates as 46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized recombinant Human TNFSF10 at 5 μg/mL (100 μL/well) can bind Human DR5, Fc Tag (Cat. No. TR2-H5255) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-7930