Learn More
ACROBiosystems Recombinant Protein;Human TRAIL R2 / DR5 / TNFRSF10B Protein;HEK293;1MG;TR2,TNFRSF10B,TRAILR2,TRAIL-R2,CD262,DR5,DR-5,KILLER,TRICK2,ZTNFR9,TRICK2A,TRICK2B,TRICKB,TRICK-2
Catalog No. 501657911
Encompass_Preferred
Recombinant Protein;Human TRAIL R2 / DR5 / TNFRSF10B Protein;1MG;Human DR5, His Tag (TR2-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession # NP_003833).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.1 kDa. The protein migrates as 15-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7911 Supplier ACROBiosystems Supplier No. TR2H52291MG
Recombinant Protein;Human TRAIL R2 / DR5 / TNFRSF10B Protein;1MG;Human DR5, His Tag (TR2-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession # NP_003833).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.1 kDa. The protein migrates as 15-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.