Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human TRAIL R2 / DR5 / TNFRSF10B Protein, Avi Tag (Avitag™);HEK293;200UG;TR2,TNFRSF10B,TRAILR2,TRAIL-R2,CD262,DR5,DR-5,KILLER,TRICK2,ZTNFR9,TRICK2A,TRICK2B,TRICKB,TRICK-2
Catalog No. 501658503
Encompass_Preferred
Recombinant Protein;Biotinylated Human TRAIL R2 / DR5 / TNFRSF10B Protein, Avi Tag (Avitag™);200UG;Biotinylated Human DR5, His Tag (TR2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession # O14763-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 16.9 kDa. The protein migrates as 22 kDa and 24 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant human TNFSF10 at 5 μg/mL (100 μL/well) can bind Biotinylated Human DR5, His Tag (Cat. No. TR2-H82E6) with a linear range of 2-156 ng/mL (QC tested).
Catalog No. 50-165-8503 Supplier ACROBiosystems Supplier No. TR2H82E6200UG
Recombinant Protein;Biotinylated Human TRAIL R2 / DR5 / TNFRSF10B Protein, Avi Tag (Avitag™);200UG;Biotinylated Human DR5, His Tag (TR2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession # O14763-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 16.9 kDa. The protein migrates as 22 kDa and 24 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant human TNFSF10 at 5 μg/mL (100 μL/well) can bind Biotinylated Human DR5, His Tag (Cat. No. TR2-H82E6) with a linear range of 2-156 ng/mL (QC tested).