Learn More
AdipoGen anti-TRAIL-R3 (human), mAb (TR3.06) (100 µg)
SDP
Supplier:  AdipoGen AG20B0029C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human TRAIL-R3. Does not cross-react with human TRAIL-R1, -R2 or -R4. Isotype: Mouse IgG1. Clone: TR3.06. Applications: FACS, IHC. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. TRAIL-R3 is a receptor for the cytotoxic ligand TRAIL. It lacks a cytoplasmic death domain and hence is not capable of inducing apoptosis. Protects cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.
Catalog No. 50-205-2932