Learn More
ACROBiosystems Recombinant Protein;Human TRAIL R4 / TNFRSF10D Protein;HEK293;100UG;TR4,TNFRSF10D,CD264,TRAILR4,DCR2,TRAIL-R4,TRUNDD,TRAIL R4
Catalog No. 501658784
Encompass_Preferred
Recombinant Protein;Human TRAIL R4 / TNFRSF10D Protein;100UG;Human CD264, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession # NP_003831).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.4 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8784 Supplier ACROBiosystems Supplier No. TR4H52H5100UG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human TRAIL R4 / TNFRSF10D Protein;100UG;Human CD264, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession # NP_003831).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.4 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.