Learn More
Aviva Systems Biology Corp TRAK1 Antibody - middle region (ARP54520_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54520_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAK1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TRAK1. Alternate names: OIP106, MILT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-935