Learn More
AdipoGen Tralomethrin
SDP
Supplier:  AdipoGen CDXT0116M100
Encompass_Preferred
Chemical. CAS 66841-25-6. Formula C22H19Br4NO3. MW 665.01. Synthetic. Potent pyrethroid insecticide. Mode of action is by modifying the gating kinetics of the sodium channels in neurons, increasing the length of time the channel remains open after a stimulus, thereby depolarizing the neuron for a longer period of time. This leads to uncontrolled spasming, paralysis and eventual death. It is effective against most members of class Arachnida as well as insects.
Catalog No. 50-256-0000