Learn More
Medchemexpress LLC HY-13519 25mg Medchemexpress, TRAM-34 CAS:289905-88-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1351925MG
Encompass_Preferred
Medchemexpress, HY-13519 25mg TRAM-34 CAS:289905-88-0 TRAM-34 is a highly selective blocker of intermediate-conductance calcium-activated K+ channel (IKCa1) (Kd=20 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9866