Learn More
TARGETMOL CHEMICALS INC TRAM-34 50MG
SDP
Catalog No. 502579497
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TRAM-34 (Triarylmethane-34) (Kd=20 nM) an effective and specific inhibitor of the intermediate-conductance Ca2+-activated K+ channel (IKCa1 KCa3.1) does not block cytochrome P450. The selective activity of TRAM-34(TRAM 34) is 200 to 1500-fold than other ion channels. purity: 99%
Catalog No. 50-257-9497 Supplier TARGETMOL CHEMICALS INC Supplier No. T618650MG
Also available in 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TRAM-34 (Triarylmethane-34) (Kd=20 nM) an effective and specific inhibitor of the intermediate-conductance Ca2+-activated K+ channel (IKCa1 KCa3.1) does not block cytochrome P450. The selective activity of TRAM-34(TRAM 34) is 200 to 1500-fold than other ion channels. purity: 99%