Learn More
Selleck Chemical LLC TRAM-34-5mg
SDP
Supplier:  Selleck Chemical LLC S11605MG
Encompass_Preferred
TRAM-34 (Triarylmethane-34) is a selective and potent inhibitor of the intermediate-conductance Ca2+-activated K+ channel (IKCa1, KCa3.1) with Kd of 20 nM, 200- to 1500-fold selective over other ion channels, and does not block cytochrome P450.
Catalog No. 50-238-9692