Learn More
ABclonal Technology TRAM1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A151421000UL
Encompass_Preferred
This gene encodes a multi-pass membrane protein that is part of the mammalian endoplasmic reticulum. The encoded protein influences glycosylation and facilitates the translocation of secretory proteins across the endoplasmic reticulum membrane by regulating which domains of the nascent polypeptide chain are visible to the cytosol during a translocational pause.
Catalog No. 50-001-04782