Learn More
Enzo Life Sciences Trametinib (10mg). CAS: 871700-17-3
SDP
Supplier:  Enzo Life Sciences ENZCHM2390010
Encompass_Preferred
Trametinib is a highly specific and potent MEK1/2 inhibitor with IC50 of 0.92 nM/1.8 nM in cell-free assays. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (20 mg/mL). Long Term Storage: -20°C.
Catalog No. 50-201-3670