Learn More
Medchemexpress LLC HY-10999 50mg Medchemexpress, Trametinib CAS:871700-17-3 Purity:>98%
SDP
Catalog No. 502028804
Encompass_Preferred
Medchemexpress, HY-10999 50mg Trametinib CAS:871700-17-3 Trametinib (GSK1120212; JTP-74057) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib activates autophagy and induces apoptosis[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8804 Supplier Medchemexpress LLC Supplier No. HY1099950MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-10999 50mg Trametinib CAS:871700-17-3 Trametinib (GSK1120212; JTP-74057) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib activates autophagy and induces apoptosis[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.