Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trametinib-d4 | 00-00-0 | 99.9% | 619.42 | C26H19D4FIN5O4 | 5 MG
SDP

Catalog No. 5000264907
Encompass_Preferred

Trametinib-d4 is the deuterium-labeled analogue of trametinib supplied as a high-purity, research-grade isotopic standard. It is used as an internal standard or reference in analytical, pharmacokinetic, and mechanistic studies of MEK inhibition and MAPK/ERK pathway modulation. The compound is provided for research use only and should be handled per laboratory safety guidance.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity (~99.9%) suitable for quantitative mass spectrometry.
  • Molecular formula and weight defined for accurate calibration and identification.
  • Soluble in DMSO for convenient preparation of stock solutions.
  • Intended for research use only; not for clinical or human use.

Catalog No. 50-002-64907 Supplier Medchemexpress LLC Supplier No. HY10999S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trametinib-d4 is the deuterium-labeled analogue of trametinib supplied as a high-purity, research-grade isotopic standard. It is used as an internal standard or reference in analytical, pharmacokinetic, and mechanistic studies of MEK inhibition and MAPK/ERK pathway modulation. The compound is provided for research use only and should be handled per laboratory safety guidance.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity (~99.9%) suitable for quantitative mass spectrometry.
  • Molecular formula and weight defined for accurate calibration and identification.
  • Soluble in DMSO for convenient preparation of stock solutions.
  • Intended for research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.