Learn More
Medchemexpress LLC Trametinib-d4 | 871700-17-3 | 99.87% | 619.42 | 1 MG
SDP
Catalog No. 5000344980
Encompass_Preferred
This product is a stable isotope-labeled compound, specifically a deuterium-labeled version of a MEK inhibitor. It is an orally active inhibitor that targets MEK1 and MEK2, demonstrating activity at very low concentrations. The compound is noted for its ability to activate autophagy and induce apoptosis. It is intended for research use only and not for human therapeutic applications.
  • Features
    • Deuterium labeled compound.
    • Targets MEK1 and MEK2 with high potency.
    • Activates autophagy.
    • Induces apoptosis.
    • Oral activity.
  • Benefits
    • Can be used as a tracer in research.
    • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
    • Deuterium substitution may influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-44980 Supplier Medchemexpress LLC Supplier No. HY10999S1MG
This product is a stable isotope-labeled compound, specifically a deuterium-labeled version of a MEK inhibitor. It is an orally active inhibitor that targets MEK1 and MEK2, demonstrating activity at very low concentrations. The compound is noted for its ability to activate autophagy and induce apoptosis. It is intended for research use only and not for human therapeutic applications.
  • Features
    • Deuterium labeled compound.
    • Targets MEK1 and MEK2 with high potency.
    • Activates autophagy.
    • Induces apoptosis.
    • Oral activity.
  • Benefits
    • Can be used as a tracer in research.
    • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
    • Deuterium substitution may influence pharmacokinetic and metabolic profiles of drugs.