Learn More
Medchemexpress LLC Trametinib-d4 | 871700-17-3 | 99.9% | 619.42 | 1 MG
SDP
Catalog No. 5000344918
Encompass_Preferred
Trametinib-d4 is the deuterium labeled Trametinib. Trametinib is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. It activates autophagy and induces apoptosis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-44918 Supplier Medchemexpress LLC Supplier No. HY10999S10MG
Trametinib-d4 is the deuterium labeled Trametinib. Trametinib is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. It activates autophagy and induces apoptosis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.