Learn More
Medchemexpress LLC Trametinib-d4 | 871700-17-3 | 99.9% | 619.42 | 25 MG
SDP
Catalog No. 5000347403
Encompass_Preferred
Trametinib-d4 is the deuterium-labeled version of Trametinib. Trametinib is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of approximately 2 nM. It also activates autophagy and induces apoptosis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution has the potential to impact the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-47403 Supplier Medchemexpress LLC Supplier No. 25MGHY10999S
Trametinib-d4 is the deuterium-labeled version of Trametinib. Trametinib is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of approximately 2 nM. It also activates autophagy and induces apoptosis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution has the potential to impact the pharmacokinetic and metabolic profiles of drugs