Learn More
Medchemexpress LLC HY-10999A 100mg , Trametinib (DMSO solvate) CAS:1187431-43-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10999A100MG
Encompass_Preferred
HY-10999A 100mg Trametinib (DMSO solvate) CAS: 1187431-43-1 Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate);JTP-74057 (DMSO solvate)) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib (DMSO solvate) activates autophagy and induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9172