Learn More
Selleck Chemical LLC Trametinib DMSO solvate S4484-1g
SDP
Catalog No. 503110478
Encompass_Preferred
Trametinib DMSO solvate is a highly specific and potent MEK1/2 inhibitor with IC50 of 0 92 nM/1 8 nM in cell-free assay Trametinib activates autophagy and induces apoptosis
Catalog No. 50-311-0478 Supplier Selleck Chemical LLC Supplier No. S44841G
Trametinib DMSO solvate is a highly specific and potent MEK1/2 inhibitor with IC50 of 0 92 nM/1 8 nM in cell-free assay Trametinib activates autophagy and induces apoptosis