Learn More
Selleck Chemical LLC Trametinib DMSO solvate-5mg
SDP
Supplier:  Selleck Chemical LLC S44845MG
Encompass_Preferred
Trametinib (GSK1120212, JTP-74057, Mekinist) DMSO solvate is a highly specific and potent MEK1/2 inhibitor with IC50 of 0.92 nM/1.8 nM in cell-free assay. Trametinib activates autophagy and induces apoptosis.
Catalog No. 50-238-9669