Learn More
Apexbio Technology LLC Tramiprosate 3687-18-1 1g
SDP
Catalog No. 502240700
Encompass_Preferred
Tramiprosate (CAS 3687-18-1) is a small molecule from the organosulfonic acid class structurally defined by the presence of a sulfonic acid functional group attached to an aliphatic backbone Tramiprosate is recognized for its interaction with amyloid-beta peptide where it inhibits aggregation and fibril formation by binding to soluble forms of amyloid-beta This mechanism has made tramiprosate a candidate for modulating amyloid pathology in neurodegenerative disease models particularly Alzheimer s disease Tramiprosate is widely utilized in biomedical research to explore the role of amyloid aggregation in disease progression and to evaluate potential therapeutic interventions
This item is not returnable. View return policy
Catalog No. 50-224-0700 Supplier Apexbio Technology LLC Supplier No. B74461000
Tramiprosate (CAS 3687-18-1) is a small molecule from the organosulfonic acid class structurally defined by the presence of a sulfonic acid functional group attached to an aliphatic backbone Tramiprosate is recognized for its interaction with amyloid-beta peptide where it inhibits aggregation and fibril formation by binding to soluble forms of amyloid-beta This mechanism has made tramiprosate a candidate for modulating amyloid pathology in neurodegenerative disease models particularly Alzheimer s disease Tramiprosate is widely utilized in biomedical research to explore the role of amyloid aggregation in disease progression and to evaluate potential therapeutic interventions