Learn More
Medchemexpress LLC Tramiprosate-d6 (homotaurine-d6) | 1131576-06-1 | 98.0% | 145.21 g/mol | C3H3D6NO3S | 5 MG
SDP
Catalog No. 5000221326
Encompass_Preferred
Tramiprosate-d6 (homotaurine-d6) is the deuterium-labeled analog of tramiprosate used as a research reagent and analytical standard. It serves as a tracer and internal standard for quantitative studies in neuroscience and analytical chemistry and is provided for research use only.
  • Deuterated analog suitable as an internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • High purity for reliable analytical results.
  • Useful for pharmacokinetic and metabolic tracer studies.
  • Provided in mg-scale quantities appropriate for analytical workflows.
Catalog No. 50-002-21326 Supplier Medchemexpress LLC Supplier No. HY14602S5MG
Tramiprosate-d6 (homotaurine-d6) is the deuterium-labeled analog of tramiprosate used as a research reagent and analytical standard. It serves as a tracer and internal standard for quantitative studies in neuroscience and analytical chemistry and is provided for research use only.
  • Deuterated analog suitable as an internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • High purity for reliable analytical results.
  • Useful for pharmacokinetic and metabolic tracer studies.
  • Provided in mg-scale quantities appropriate for analytical workflows.