Learn More
Medchemexpress LLC Tramiprosate-d6 (Homotaurine-d6) | 1131576-06-1 | 98.0% | C3H3D6NO3S | 1 MG
SDP
Catalog No. 5000355279
Encompass_Preferred
Tramiprosate-d6 is the deuterium labeled Tramiprosate. It is an orally active and brain-penetrant natural amino acid found in various species of red marine algae. It binds to soluble Aβ and maintains Aβ in a non-fibrillar form. Tramiprosate also acts as a GABA analog, offering neuroprotection, anticonvulsion, and antihypertension effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-55279 Supplier Medchemexpress LLC Supplier No. HY14602S1MG
Tramiprosate-d6 is the deuterium labeled Tramiprosate. It is an orally active and brain-penetrant natural amino acid found in various species of red marine algae. It binds to soluble Aβ and maintains Aβ in a non-fibrillar form. Tramiprosate also acts as a GABA analog, offering neuroprotection, anticonvulsion, and antihypertension effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.