Learn More
Medchemexpress LLC Tramiprosate-d6 (homotaurine-d6) | 1131576-06-1 | 98.0% | C3H3D6NO3S | 10MG
SDP
Catalog No. 5000259998
Encompass_Preferred
Tramiprosate-d6 (homotaurine-d6) is the deuterium-labeled analogue of tramiprosate intended for research and analytical applications. It is used as a tracer or internal standard for quantitative methods and supports studies of amyloid-β interactions, neurochemical analysis, and method validation.
Catalog No. 50-002-59998 Supplier Medchemexpress LLC Supplier No. HY14602S10MG
Tramiprosate-d6 (homotaurine-d6) is the deuterium-labeled analogue of tramiprosate intended for research and analytical applications. It is used as a tracer or internal standard for quantitative methods and supports studies of amyloid-β interactions, neurochemical analysis, and method validation.