Learn More
Description
- (L-Methionine, [35S]: L-Cysteine, [35S])
- Sp. Act. >1000 Ci/mmol; >37.0 TBq/mmol
- Net L-Methionine Radioactivity 1 mCi, 5 mCi and 10 mCi, respectively
- Metabolic labeling reagent derived from [35S]E. coli hydrolysate containing >70% L-methionine, [35S], ∼15% L-cysteine, [35S]
- Solution in 50 mM L-lysine, pH 7.4
- Containing 10 mM 2-mercaptoethanol
- Concentration ∼10 mCi/ml; ∼370 MBq/ml
- Packaged under nitrogen in the Versatainer™Packaging System
- For Higher Incorporation in Metabolic Labeling:
- MP CELLect™ Met/Cys Deficient Media
- ∙ Deficient in Methionine
- Allows methionine incorporation in metabolic labeling
- Deficient in Cysteine
Optimal media for labeling with 35S-Methionine since cysteine is a metabolic precursor to methionine. Also allows for 35S-Cysteine incorporation which is of interest to users of Tran35S-label and 35S-Cysteine
Specifications
Specifications
| Quantity | 1.4 mci |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.