Learn More
Medchemexpress LLC HY-B0592 10mg Medchemexpress, Trandolapril CAS:87679-37-6 Purity:>98%
SDP
Catalog No. 502082614
Encompass_Preferred
Medchemexpress, HY-B0592 10mg Trandolapril CAS:87679-37-6 Trandolapril (RU44570) is a nonsulfhydryl prodrug that is hydrolysed to the active diacid Trandolaprilat. Trandolapril is an orally administered angiotensin converting enzyme (ACE) inhibitor that has been used in the treatment of hypertension and congestive heart failure (CHF), and after myocardial infarction (MI). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2614 Supplier Medchemexpress LLC Supplier No. HYB059210MG
Medchemexpress, HY-B0592 10mg Trandolapril CAS:87679-37-6 Trandolapril (RU44570) is a nonsulfhydryl prodrug that is hydrolysed to the active diacid Trandolaprilat. Trandolapril is an orally administered angiotensin converting enzyme (ACE) inhibitor that has been used in the treatment of hypertension and congestive heart failure (CHF), and after myocardial infarction (MI). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.