Learn More
Medchemexpress LLC Trandolaprilate hydrate | 951393-55-8 | 97.3% | C22H32N2O6 | 1 MG
SDP
Catalog No. 5000433387
Encompass_Preferred
Trandolaprilate hydrate is a potent angiotensin-converting enzyme (ACE) inhibitor that partially inhibits angiotensin-I-mediated c-fos induction. It is the main bioactive metabolite of Trandolapril and exhibits high lipophilicity. For storage, the product should be kept at -20°C, sealed, away from moisture. If stored in solvent, it is stable at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).
  • Potent angiotensin-converting enzyme (ACE) inhibitor.
  • Partially inhibits angiotensin-I-mediated c-fos induction.
  • Bioactive metabolite.
  • High lipophilicity.
Catalog No. 50-004-33387 Supplier Medchemexpress LLC Supplier No. HYA0116A1MG
Trandolaprilate hydrate is a potent angiotensin-converting enzyme (ACE) inhibitor that partially inhibits angiotensin-I-mediated c-fos induction. It is the main bioactive metabolite of Trandolapril and exhibits high lipophilicity. For storage, the product should be kept at -20°C, sealed, away from moisture. If stored in solvent, it is stable at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).
  • Potent angiotensin-converting enzyme (ACE) inhibitor.
  • Partially inhibits angiotensin-I-mediated c-fos induction.
  • Bioactive metabolite.
  • High lipophilicity.