Learn More
Lkt Laboratories CAS 1197-18-8, ≥98%, Plasminogen inhibitor used to decrease blood loss in cardiac surgery and trauma.
SDP
Supplier:  Lkt Laboratories T6811
Encompass_Preferred
Tranexamic acid is clinically used to decrease blood loss in cardiac surgery and trauma; it inhibits plasmin/plasminogen. In vivo, tranexamic acid inhibits fibrinolysis, decreases bleeding time, and increases thrombus formation. Additionally, tranexamic acid accelerates skin barrier recovery and upregulates occluding expression in UV-damaged skin.
Catalog No. NC2522903