Learn More
Apexbio Technology LLC Tranexamic Acid, 1197-18-8, MFCD00001466, 10 g
SDP
Supplier:  Apexbio Technology LLC B185810000
Encompass_Preferred
MF: C8H15NO2, MW: 157.21, Solubility: 6.6mg/mL in H2O. Tranexamic Acid is an antifibrinolytic, blocking lysine-binding sites (LBS) of plasmin and elastase-derived plasminogen fragments, with IC50 value of 5 mM.
Catalog No. 50-101-0014