Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tranexamic acid | 1197-18-8 | 10 MG
SDP

Catalog No. 5000298248
Encompass_Preferred

Tranexamic acid is an antifibrinolytic compound supplied as an analytical standard for research and analytical applications. It is used in studies of hemostasis, plasminogen activation, and related biochemical assays.

  • Analytical standard intended for research and analytical use.
  • Antifibrinolytic agent; cyclic lysine analog.
  • CAS number 1197-18-8.
  • Molecular weight 157.21 g/mol.
  • Chemical formula C8H15NO2.
  • Available as a 10 MG package.
  • Purity not specified on the datasheet.

Catalog No. 50-002-98248 Supplier Medchemexpress LLC Supplier No. HYB0149R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tranexamic acid is an antifibrinolytic compound supplied as an analytical standard for research and analytical applications. It is used in studies of hemostasis, plasminogen activation, and related biochemical assays.

  • Analytical standard intended for research and analytical use.
  • Antifibrinolytic agent; cyclic lysine analog.
  • CAS number 1197-18-8.
  • Molecular weight 157.21 g/mol.
  • Chemical formula C8H15NO2.
  • Available as a 10 MG package.
  • Purity not specified on the datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.