Learn More
Medchemexpress LLC Tranexamic acid | 1197-18-8 | 99.88% | 500 MG
SDP
Catalog No. 5000450516
Encompass_Preferred
Tranexamic acid, a cyclic analog of lysine, is an orally active antifibrinolytic agent that functions as a hemostatic agent. It attenuates the effects of severe trauma, inhibits urokinase plasminogen activator, and can ameliorate dry wrinkles. This compound is used in research for hemostasis studies and as a PROTAC linker.
  • Attenuates effects of severe trauma by enhancing mitochondrial respiration, suppressing damage-associated molecular patterns (DAMPs) release, and improving capillary integrity.
  • Inhibits cell migration in MDA-MB-231 breast cancer cells.
  • Facilitates immune activation after traumatic brain injury in plasminogen-deficient mice.
  • Decreases mast cell proliferation and increases fibroblast proliferation, ameliorating wrinkles caused by skin dryness in mice.
  • Utilized in research for hemostasis studies.
  • Serves as a PROTAC linker to synthesize PROTACs.
Catalog No. 50-004-50516 Supplier Medchemexpress LLC Supplier No. HYB0149500MG
Tranexamic acid, a cyclic analog of lysine, is an orally active antifibrinolytic agent that functions as a hemostatic agent. It attenuates the effects of severe trauma, inhibits urokinase plasminogen activator, and can ameliorate dry wrinkles. This compound is used in research for hemostasis studies and as a PROTAC linker.
  • Attenuates effects of severe trauma by enhancing mitochondrial respiration, suppressing damage-associated molecular patterns (DAMPs) release, and improving capillary integrity.
  • Inhibits cell migration in MDA-MB-231 breast cancer cells.
  • Facilitates immune activation after traumatic brain injury in plasminogen-deficient mice.
  • Decreases mast cell proliferation and increases fibroblast proliferation, ameliorating wrinkles caused by skin dryness in mice.
  • Utilized in research for hemostasis studies.
  • Serves as a PROTAC linker to synthesize PROTACs.