Learn More
Medchemexpress LLC Tranexamic acid | 1197-18-8 | 99.9% | 250 MG
SDP
Catalog No. 5000405576
Encompass_Preferred
Tranexamic acid (Standard) is the analytical standard of Tranexamic acid, intended for research and analytical applications. It is a cyclic analog of lysine and an orally active antifibrinolytic agent. The compound is used for research of hemostasis.
  • Analytical standard for tranexamic acid.
  • Intended for research and analytical applications.
  • Attenuates the effects of severe trauma.
  • Inhibits urokinase plasminogen activator.
  • Ameliorates dry wrinkles.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-05576 Supplier Medchemexpress LLC Supplier No. 250MGHYB0149R
Tranexamic acid (Standard) is the analytical standard of Tranexamic acid, intended for research and analytical applications. It is a cyclic analog of lysine and an orally active antifibrinolytic agent. The compound is used for research of hemostasis.
  • Analytical standard for tranexamic acid.
  • Intended for research and analytical applications.
  • Attenuates the effects of severe trauma.
  • Inhibits urokinase plasminogen activator.
  • Ameliorates dry wrinkles.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.