Learn More
Medchemexpress LLC HY-B0149 10g Medchemexpress, Tranexamic acid CAS:1197-18-8 Purity:98%
SDP
Catalog No. 501639209
Encompass_Preferred
Medchemexpress,HY-B0149 10g Tranexamic acid CAS:1197-18-8 Formula:C8H15NO2 Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9209 Supplier Medchemexpress LLC Supplier No. HYB014910G
Medchemexpress,HY-B0149 10g Tranexamic acid CAS:1197-18-8 Formula:C8H15NO2 Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.