Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tranexamic acid-13C2,15N | 1292837-95-6 | 1 MG
SDP

Catalog No. 5000405346
Encompass_Preferred

Tranexamic acid-13C2,15N is a stable isotope-labeled form of Tranexamic acid. This antifibrinolytic agent has been shown to alleviate liver damage and fibrosis in mouse models of chronic bile duct injury. It is intended for research use only and not for sale to patients.

  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight: 160.19
  • Formula: C₆¹³C₂H₁₅¹⁵NO₂

Catalog No. 50-004-05346 Supplier Medchemexpress LLC Supplier No. HYB0149S31MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tranexamic acid-13C2,15N is a stable isotope-labeled form of Tranexamic acid. This antifibrinolytic agent has been shown to alleviate liver damage and fibrosis in mouse models of chronic bile duct injury. It is intended for research use only and not for sale to patients.

  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight: 160.19
  • Formula: C₆¹³C₂H₁₅¹⁵NO₂

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.