Learn More
Medchemexpress LLC Tranexamic acid-d2-1 | 2714435-89-7 | 99.2% | 500 UG
SDP
Catalog No. 5000406306
Encompass_Preferred
Tranexamic acid-d2-1 is the deuterium labeled Tranexamic acid. Tranexamic acid (Transamin) is an antifibrinolytic that blocks lysine-binding sites of plasmin and elastase-derived plasminogen fragments with an IC50 of 5 mM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06306 Supplier Medchemexpress LLC Supplier No. HYB0149S1500UG
Tranexamic acid-d2-1 is the deuterium labeled Tranexamic acid. Tranexamic acid (Transamin) is an antifibrinolytic that blocks lysine-binding sites of plasmin and elastase-derived plasminogen fragments with an IC50 of 5 mM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.