Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tranexamic acid impurity 1 | 1197-17-7 | MFCD19706018 | ≥98.0% | 157.21 g/mol | C8H15NO2 | 250 MG
SDP

Catalog No. 5000209194
Encompass_Preferred

Tranexamic acid impurity 1 is a characterized impurity of the active pharmaceutical ingredient tranexamic acid, supplied for research and analytical purposes as a drug intermediate or impurity reference. The substance is provided with a certificate of analysis and recommended storage instructions for laboratory use.

  • Used as an analytical reference or impurity standard.
  • Purity: ≥98.0%.
  • Chemical formula: C8H15NO2; molecular weight: 157.21 g/mol.
  • Supplied in multiple pack sizes (e.g., 250 MG) for flexibility in experiments.
  • Intended for research use only; not for clinical or diagnostic use.

Catalog No. 50-002-09194 Supplier Medchemexpress LLC Supplier No. 250MGHYW063191
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tranexamic acid impurity 1 is a characterized impurity of the active pharmaceutical ingredient tranexamic acid, supplied for research and analytical purposes as a drug intermediate or impurity reference. The substance is provided with a certificate of analysis and recommended storage instructions for laboratory use.

  • Used as an analytical reference or impurity standard.
  • Purity: ≥98.0%.
  • Chemical formula: C8H15NO2; molecular weight: 157.21 g/mol.
  • Supplied in multiple pack sizes (e.g., 250 MG) for flexibility in experiments.
  • Intended for research use only; not for clinical or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.